Abnova
Manufacturer Code:H00010005K
Catalog # 89-201-493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ACOT8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ACOT8 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Acyl-CoA thioesterase 8; Acyl-coenzyme A thioesterase 8; choloyl-CoA hydrolase; Choloyl-coenzyme A thioesterase; hACTE-III; HIV-Nef associated acyl-CoA thioesterase; HIV-Nef-associated acyl-CoA thioesterase; HNAACTE PTE-2 PTE1 PTE2 hACTE-III hTE; long-chain fatty-acyl-CoA hydrolase; Nef (lentivirus myristoylated factor) associated protein 1; palmitoyl-CoA hydrolase; peroxisomal acyl-CoA thioesterase 1; Peroxisomal acyl-CoA thioesterase 2; Peroxisomal acyl-coenzyme A thioester hydrolase 1; Peroxisomal long-chain acyl-CoA thioesterase 1; PTE-1; PTE-2; Thioesterase II; thioesterase III
Gene Aliases: ACOT8; ACTEIII; hACTE-III; HNAACTE; hTE; NAP1; PTE-1; PTE-2; PTE1; PTE2
UniProt ID: (Human) O14734
Entrez Gene ID: (Human) 10005
Molecular Function:
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.