Novus Biologicals
Manufacturer Code:NBP222569JF646
Catalog # NBP22569646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D5 |
Immunogen | Recombinant human ACOT7 (1-370aa) purified from E. coli |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ACH1 acyl-CoA thioesterase 7CTE-IIa BACHEC 3.1.2.2 brain acyl CoA hydrolase Brain acyl-CoA hydrolase CTE-IILACH cytosolic acyl coenzyme A thioester hydrolase hBACH long chain Long chain acyl-CoA thioester hydrolase; acyl-CoA thioesterase 2; Acyl-CoA thioesterase 7; acyl-CoA thioesterase, long chain; BACH; brain acyl CoA hydrolase; Brain acyl-CoA hydrolase; CTE-II; CTE-IIa; Cytosolic acyl coenzyme A thioester hydrolase; Long chain acyl-CoA thioester hydrolase
Gene Aliases: ACH1; ACOT7; ACT; BACH; CTE-II; hBACH; LACH; LACH1
UniProt ID: (Human) O00154
Entrez Gene ID: (Human) 11332
Molecular Function:
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.