Proteintech
Manufacturer Code:159721AP150UL
Catalog # 50-173-0611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACOT7 Fusion Protein Ag8759 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 50-kDa brain acyl-CoA hydrolase; ACH1; ACH1 ACOT7 ACT acyl CoA thioesterase 7 BACH Brain acyl CoA hydrolase CTE II CTE IIa hBACH LACH LACH1; ACT; acyl-CoA thioesterase 2; Acyl-CoA thioesterase 7; acyl-CoA thioesterase, long chain; BACH; brain acyl CoA hydrolase; Brain acyl-CoA hydrolase; CTE-II; CTE-IIa; CTE-IIb; Cytosolic acyl coenzyme A thioester hydrolase; LACH1; Long chain acyl-CoA thioester hydrolase; MTE-II
Gene Aliases: 2410041A17Rik; ACH1; ACOT7; ACT; BACH; CTE-II; CTE-IIa; hBACH; LACH; LACH1
UniProt ID: (Human) O00154, (Mouse) Q91V12, (Rat) Q64559
Entrez Gene ID: (Human) 11332, (Mouse) 70025, (Rat) 26759
Molecular Function:
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.