Invitrogen
Manufacturer Code:PA570857
Catalog # PIPA570857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human ACOT6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acyl-CoA thioesterase 6; acyl-coenzyme A thioesterase 6; Putative acyl-CoA thioesterase 6; Putative acyl-coenzyme A thioesterase 6
Gene Aliases: ACOT6; c14_5530; C14orf42; RGD1309669
UniProt ID: (Human) Q3I5F7
Entrez Gene ID: (Human) 641372, (Pig) 100152868, (Horse) 100056126, (Rat) 299193
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.