Novus Biologicals
Manufacturer Code:NBP2825790.1ML
Catalog # NB030508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human ACOT6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acyl-CoA thioesterase 6c14_5530 C14orf42 chromosome 14 open reading frame 42 EC 3.1.2.- EC 3.1.2.2 MGC161656 MGC161658 putative acyl-coenzyme A thioesterase 6; Putative acyl-CoA thioesterase 6; Putative acyl-coenzyme A thioesterase 6
Gene Aliases: ACOT6; c14_5530; C14orf42
UniProt ID: (Human) Q3I5F7
Entrez Gene ID: (Human) 641372
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.