Proteintech
Manufacturer Code:229361AP150UL
Catalog # 50-173-3967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACOT6 Fusion Protein Ag18981 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACOT6 acyl CoA thioesterase 6 c14&lowbar5530 C14orf42; Acyl-CoA thioesterase 6; Acyl-coenzyme A thioesterase 6; Putative acyl-CoA thioesterase 6; Putative acyl-coenzyme A thioesterase 6
Gene Aliases: 4632408A20Rik; A330054B07; ACOT6; BE688602; c14_5530; C14orf42; RGD1309669
UniProt ID: (Human) Q3I5F7, (Mouse) Q32Q92
Entrez Gene ID: (Human) 641372, (Rat) 299193, (Mouse) 217700
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.