Novus Biologicals
Manufacturer Code:NBP2825750.1ML
Catalog # NB030504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human ACOT4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA thioesterase 4; acyl-CoA thioesterase 4EC 3.1.2.2 acyl-coenzyme A thioesterase 4 Peroxisomal acyl coenzyme A thioester hydrolase Ib peroxisomal acyl-CoA thioesterase 2B Peroxisomal long-chain acyl-CoA thioesterase Ib PTE-2b PTE2BPTE1B PTEIB PTE-IbFLJ31235; Acyl-coenzyme A thioesterase 4; Peroxisomal acyl coenzyme A thioester hydrolase Ib; peroxisomal acyl-CoA thioesterase 2B; Peroxisomal long-chain acyl-CoA thioesterase Ib; Peroxisomal succinyl-coenzyme A thioesterase; PTE-2b; PTE-Ib
Gene Aliases: ACOT4; PTE-Ib; PTE1B; PTE2B; PTEIB
UniProt ID: (Human) Q8N9L9
Entrez Gene ID: (Human) 122970
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.