Novus Biologicals
Manufacturer Code:NBP272389F
Catalog # NBP272389F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A11 |
Immunogen | Full length recombinant protein of human ACOT12 (NP_570123) produced in HEK293T cells. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Acetyl-coenzyme A thioesterase; Acyl-CoA thioester hydrolase 12; Acyl-CoA thioester hydrolase 12 acyl-CoA thioesterase 12EC 3.1.2.1 acyl-coenzyme A thioesterase 12 Cach CACH1 CACH-1 Cytoplasmic acetyl-CoA hydrolase 1 cytosolic acetyl-CoA hydrolase hCACH-1 StARD15 STARD15MGC105114 StAR-related lipid transfer (START) domain containing 15 START domain-containing protein 15 THEAL; Acyl-CoA thioesterase 12; Acyl-coenzyme A thioesterase 12; CACH-1; Cytoplasmic acetyl-CoA hydrolase 1; cytosolic acetyl-CoA hydrolase; hCACH-1; StAR-related lipid transfer (START) domain containing 15; StARD15; START domain-containing protein 15
Gene Aliases: ACOT12; CACH; CACH-1; CACH1; STARD15; THEAL
UniProt ID: (Human) Q8WYK0
Entrez Gene ID: (Human) 134526
Molecular Function:
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.