Invitrogen
Manufacturer Code:PA531981
Catalog # PIPA531981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 290 and 537 of Human ACOT12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetyl-coenzyme A thioesterase; Acyl-CoA thioester hydrolase 12; Acyl-CoA thioesterase 12; Acyl-coenzyme A thioesterase 12; CACH-1; CACH-1 Cach STARD15 THEAL START domain-containing protein 15 StAR-related lipid transfer (START) domain containing 15 acyl-CoA thioester hydrolase 12 acyl-coenzyme A thioesterase 12 cytoplasmic acetyl-CoA hydrolase 1 cytosolic acetyl-CoA hydrolase hCACH-1; Cytoplasmic acetyl-CoA hydrolase 1; cytosolic acetyl-CoA hydrolase; hCACH-1; StAR-related lipid transfer (START) domain containing 15; StARD15; START domain-containing protein 15
Gene Aliases: ACOT12; CACH; CACH-1; CACH1; STARD15; THEAL
UniProt ID: (Human) Q8WYK0
Entrez Gene ID: (Human) 134526
Molecular Function:
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.