Novus Biologicals
Manufacturer Code:NBP104289FR
Catalog # NBP104289FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | J4B2 |
Immunogen | Recombinant human ACOT11 (19-250aa) purified from E. coli |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA thioester hydrolase 11; Acyl-CoA thioester hydrolase 11 acyl-CoA thioesterase 11BFIT1 Adipose-associated thioesterase BFIT2 BFITDKFZp667O1916 Brown fat-inducible thioesterase EC 3.1.2.- EC 3.1.2.1 KIAA0707acyl-coenzyme A thioesterase 11 STARD14 START domain containing 14 THEAStAR-related lipid transfer (START) domain containing 14 THEM1 thioesterase superfamily member 1 thioesterase adipose associated; Acyl-CoA thioesterase 11; Acyl-coenzyme A thioesterase 11; Adipose-associated thioesterase; BFIT; Brown fat-inducible thioesterase; Palmitoyl-coenzyme A thioesterase; StAR-related lipid transfer (START) domain containing 14; START domain containing 14; thioesterase superfamily member 1; thioesterase, adipose associated
Gene Aliases: ACOT11; BFIT; KIAA0707; STARD14; THEA; THEM1
UniProt ID: (Human) Q8WXI4
Entrez Gene ID: (Human) 26027
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.