Abnova
Manufacturer Code:PAB18049
Catalog # 89-119-625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human ACO1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ACO1.
For Research Use Only
Protein Aliases: Aconitase; aconitase 1, soluble; aconitate hydratase, cytoplasmic; ACONS; Citrate hydro-lyase; Cytoplasmic aconitate hydratase; cytosplasmic aconitase; epididymis luminal protein 60; Ferritin repressor protein; IRE-BP 1; IREB1; IREBP; IREBP1; Iron regulatory protein 1; iron-responsive element binding protein 1; Iron-responsive element-binding protein 1; IRP1; soluble aconitase
Gene Aliases: ACO1; ACONS; HEL60; IREB1; IREBP; IREBP1; IRP1
UniProt ID: (Human) P21399
Entrez Gene ID: (Human) 48
Molecular Function:
dehydratase
hydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.