Invitrogen
Manufacturer Code:PA5113272
Catalog # PIPA5113272
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human 2-amino-3-carboxymuconate-6-semialdehyde decarboxylase protein (33-129AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-amino-3-carboxylmuconate-6-semialdehyde decarboxylase 2-amino-3-carboxymuconate-6-semialdehyde decarboxylase 2-amino-3-carboxymuconate-6-semialdehyde decarboxylase-like Acmsd amino carboxymuconate semialdehyde decarboxylase aminocarboxymuconate semialdehyde decarboxylase BOS_1852 LOC100154768 LOC100350080 Picolinate carboxylase zgc:162888; 2-amino-3-carboxymuconate-6-semialdehyde decarboxylase; Picolinate carboxylase
Gene Aliases: ACMSD
UniProt ID: (Human) Q8TDX5, (Rat) Q8R5M5
Entrez Gene ID: (Human) 130013, (Rat) 171385
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.