Invitrogen
Manufacturer Code:PA528389
Catalog # PIPA528389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 25 and 313 of Human ACK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACK ACK-1 ACK1 p21cdc42Hs activated CDC42 kinase 1 activated Cdc42-associated kinase 1 activated p21cdc42Hs kinase tyrosine kinase non-receptor protein 2; ACK-1; Activated CDC42 kinase 1; activated Cdc42-associated kinase 1; activated p21cdc42Hs kinase; Cdc42 GTPase-inhibiting protein; Non-receptor protein tyrosine kinase Ack; P21cdc42Hs kinase; Tyrosine kinase non-receptor protein 2; tyrosine kinase, non-receptor, 2
Gene Aliases: ACK; ACK-1; ACK1; Cdgip; p21cdc42Hs; Pyk1; TNK2
UniProt ID: (Human) Q07912, (Mouse) O54967
Entrez Gene ID: (Human) 10188, (Mouse) 51789
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.