Invitrogen
Manufacturer Code:PA514764
Catalog # PIPA514764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 10-39 amino acids from the N-terminal region of human ACK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACK-1; Activated CDC42 kinase 1; activated Cdc42-associated kinase 1; Activated p21cdc42Hs kinase; Tyrosine kinase non-receptor protein 2; tyrosine kinase, non-receptor, 2
Gene Aliases: ACK; ACK-1; ACK1; p21cdc42Hs; TNK2
UniProt ID: (Human) Q07912
Entrez Gene ID: (Human) 10188
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.