Abnova
Manufacturer Code:H00000043K
Catalog # 89-206-324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ACHE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ACHE peptide using ARM Technology.
For Research Use Only
Protein Aliases: Acetylcholinesterase; acetylcholinesterase (Yt blood group); AChE; apoptosis-related acetylcholinesterase; ARACHE N-ACHE YT; Yt blood group
Gene Aliases: ACEE; ACHE; ARACHE; N-ACHE; YT
UniProt ID: (Human) P22303
Entrez Gene ID: (Human) 43
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.