Invitrogen
Manufacturer Code:PA560608
Catalog # PIPA560608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ACF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACF ACF64 ACF65 APOBEC1CF ASP APOBEC-1 stimulating protein apo-B RNA editing protein apobec-1 complementation factor (ACF) (ASP); apo-B RNA editing protein; apobec-1 complementation factor (ACF) (ASP); APOBEC-1 stimulating protein; APOBEC1 complementation factor; APOBEC1-stimulating protein
Gene Aliases: A1CF; ACF; ACF64; ACF65; APOBEC1CF; ASP
UniProt ID: (Human) Q9NQ93
Entrez Gene ID: (Human) 29974
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.