Invitrogen
Manufacturer Code:PA541849
Catalog # PIPA541849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human A1CF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acf; apo-B RNA editing protein; apobec-1 complementation factor; apobec-1 complementation factor (ACF) (ASP); Apobec-1 complementation factor APOBEC-1 stimulating protein; Apobec-1 complementation factor, APOBEC-1 stimulating protein; APOBEC-1 stimulating protein; APOBEC1 complementation factor; APOBEC1 complementation factor ACF ACF64 ACF65 APOBEC1CF ASP MGC163391 RP11-564C4.2; APOBEC1 complementation factort; APOBEC1-stimulating protein
Gene Aliases: 1810073H04Rik; A1CF; A1cft; ACF; ACF64; ACF65; Apobec-1; APOBEC1CF; ASP
UniProt ID: (Human) Q9NQ93, (Mouse) Q5YD48, (Rat) Q923K9
Entrez Gene ID: (Dog) 477581, (Rabbit) 100339264, (Human) 29974, (Horse) 100071844, (Guinea pig) 100730935, (Mouse) 69865, (Rat) 170912, (Zebrafish) 562916
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.