Invitrogen
Manufacturer Code:PA575832
Catalog # PIPA575832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human ACER2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 3-like; alkaline CDase 2; Alkaline ceramidase 2; Alkaline ceramidase 2 AlkCDase 2 Alkaline CDase 2 haCER2 Acylsphingosine deacylase 3-like N-acylsphingosine amidohydrolase 3-like ACER2 ASAH3L PP11646; AlkCDase 2; ceramide hydrolase; haCER2; N-acylsphingosine amidohydrolase 3-like
Gene Aliases: ACER2; ALKCDase2; ASAH3L; PP11646
UniProt ID: (Human) Q5QJU3
Entrez Gene ID: (Human) 340485, (Rat) 313339
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.