Invitrogen
Manufacturer Code:PA539016
Catalog # PIPA539016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human ASAH3L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 3-like; alkaline CDase 2; Alkaline ceramidase 2; AlkCDase 2; ASAH3L; cancer related gene-liver 1 (CRG-L1); Cancer-related gene liver 1 protein; ceramide hydrolase; CRG-L1; haCER2; N-acylsphingosine amidohydrolase 3-like
Gene Aliases: 2410116I05Rik; ACER2; ALKCDase2; ASAH3L; CRG-L1; maCER2; PP11646
UniProt ID: (Human) Q5QJU3, (Mouse) Q8VD53
Entrez Gene ID: (Human) 340485, (Mouse) 230379
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.