Invitrogen
Manufacturer Code:PA5114249
Catalog # PIPA5114249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of Human ACER2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410116I05Rik ACER2 Acylsphingosine deacylase 3-like alkaline CDase 2 Alkaline ceramidase 2 AlkCDase 2 ALKCDase2 ASAH3L cancer related gene-liver 1 (CRG-L1) Cancer-related gene liver 1 protein ceramide hydrolase CRG-L1 haCER2 maCER2 N-acylsphingosine amidohydrolase 3-like PP11646; Acylsphingosine deacylase 3-like; alkaline CDase 2; Alkaline ceramidase 2; AlkCDase 2; ceramide hydrolase; haCER2; N-acylsphingosine amidohydrolase 3-like
Gene Aliases: ACER2; ALKCDase2; ASAH3L; PP11646
UniProt ID: (Human) Q5QJU3
Entrez Gene ID: (Human) 340485
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.