OriGene
Manufacturer Code:CF803841
Catalog # 50-166-6694
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 18-237 of human ACE2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACE-related carboxypeptidase; ACEH; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; Angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; Angiotensin-converting enzyme-related carboxypeptidase; Metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2
Gene Aliases: ACE2; ACEH; UNQ868/PRO1885
UniProt ID: (Human) Q9BYF1
Entrez Gene ID: (Human) 59272
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.