Invitrogen
Manufacturer Code:PA547488
Catalog # PIPA547488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit mouse ACE-2 Gln18-Thr740 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACE-related carboxypeptidase; angiotensin I converting enzyme 2; Angiotensin-converting enzyme 2; Processed angiotensin-converting enzyme 2; UNQ868/PRO1885 ACEH ACE-related carboxypeptidase angiotensin I converting enzyme 2 angiotensin-converting enzyme 2 angiotensin-converting enzyme homolog metalloprotease MPROT15
Gene Aliases: 2010305L05Rik; Ace2
UniProt ID: (Mouse) Q8R0I0
Entrez Gene ID: (Mouse) 70008
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.