R&D Systems
Manufacturer Code:AF929SP
Catalog # AF929SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ACE/CD143 Leu30-Leu1261 Accession # P12821 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACE; ACE1angiotensin converting enzyme somatic isoform angiotensin I converting enzyme (peptidyl-dipeptidase A) 1 carboxycathepsin CD143 CD143 antigen DCP DCP1 DCP1angiotensin-converting enzyme dipeptidyl carboxypeptidase 1 Dipeptidyl carboxypeptidase I EC 3.2.1.- EC 3.4.15.1 Kininase II MGC26566 MVCD3 peptidase P testicular ECA; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; Angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; carboxycathepsin; CD143; CD143 antigen; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase I; Kininase II; peptidase P; testicular ECA
Gene Aliases: ACE; ACE1; CD143; DCP; DCP1; ICH; MVCD3
UniProt ID: (Human) P12821
Entrez Gene ID: (Human) 1636
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.