Invitrogen
Manufacturer Code:MA535099
Catalog # PIMA535099
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0577 |
Immunogen | A synthesized peptide derived from human ACE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ACE; ACE ACE1 Angiotensin-converting enzyme Angiotensin-converting enzyme soluble form AW208573 BOS_19238 CD143 DCP DCP1 Dipeptidyl carboxypeptidase I I79_009670 ICH Kininase II MGC26566 MVCD3 StsRR92; angiotensin 1 converting enzyme 1; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1); Angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; carboxycathepsin; CD143; CD143 antigen; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme); Dipeptidyl carboxypeptidase I; dipeptidyl peptidase; Kininase II; peptidase P; testicular ECA
Gene Aliases: ACE; ACE1; AW208573; CD143; DCP; DCP1; ICH; MVCD3; StsRR92
UniProt ID: (Human) P12821, (Mouse) P09470, (Rat) P47820
Entrez Gene ID: (Human) 1636, (Mouse) 11421, (Rat) 24310
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.