Invitrogen
Manufacturer Code:MA532956
Catalog # PIMA532956
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6C3 |
Immunogen | Synthetic peptide corresponding to the N-terminus of Human ACE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACE; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; Angiotensin-converting enzyme; Angiotensin-converting enzyme Short name&equalsACE Dipeptidyl carboxypeptidase I CD143 ACE1 DCP DCP1 Angiotensin Converting Enzyme 1; Angiotensin-converting enzyme, soluble form; carboxycathepsin; CD143; CD143 antigen; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase I; dipeptidyl peptidase; Kininase II; peptidase P; testicular ECA
Gene Aliases: ACE; ACE1; AW208573; CD143; DCP; DCP1; ICH; MVCD3
UniProt ID: (Human) P12821, (Mouse) P09470
Entrez Gene ID: (Human) 1636, (Mouse) 11421
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.