Invitrogen
Manufacturer Code:MA537594
Catalog # PIMA537594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 536CT24.1.3 |
Immunogen | KLH conjugated synthetic peptide between 1274-1306 amino acids from human ACE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACE; ACE ACE1 angiotensin 1 converting enzyme 1 angiotensin converting enzyme somatic isoform angiotensin I converting enzyme angiotensin I converting enzyme (peptidyl-dipeptidase A) 1 angiotensin I converting enzyme 1 angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1) angiotensin-converting enzyme Angiotensin-converting enzyme soluble form AW208573 carboxycathepsin CD143 CD143 antigen DCP Dcp1 dipeptidyl carboxypeptidase 1 Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme) dipeptidyl carboxypeptidase I dipeptidyl peptidase ICH kininase II MGC26566 MVCD3 peptidase P StsRR92 testicular ECA; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; Angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; carboxycathepsin; CD143; CD143 antigen; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase I; dipeptidyl peptidase; Kininase II; peptidase P; testicular ECA
Gene Aliases: ACE; ACE1; AW208573; CD143; DCP; DCP1; ICH; MVCD3
UniProt ID: (Human) P12821, (Mouse) P09470
Entrez Gene ID: (Human) 1636, (Mouse) 11421
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.