Invitrogen
Manufacturer Code:PA585661
Catalog # PIPA585661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human ACE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACE; Angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; CD143; CD143 Antibody DCP1 Antibody angiotensin I converting enzyme Antibody ACE1 Antibody DCP Antibody ICH Antibody MVCD3 Antibody; Dipeptidyl carboxypeptidase I; dipeptidyl peptidase; Kininase II
Gene Aliases: Ace; AW208573; CD143; Dcp1
UniProt ID: (Mouse) P09470
Entrez Gene ID: (Mouse) 11421
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.