Invitrogen
Manufacturer Code:PA585115
Catalog # PIPA585115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human ACE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACE; ACE1 antibody CD143 antibody DCP antibody DCP1 antibody MGC26566 antibody MVCD3 antibody ACE antibody dipeptidyl carboxypeptidase 1 antibody angiotensin-converting enzyme antibody carboxycathepsin antibody peptidase P antibody dipeptidyl carboxypeptidase I antibody kininase II antibody angiotensin converting enzyme somatic isoform antibody CD143 antigen antibody testicular ECA antibody angiotensin I converting enzyme (peptidyl-dipeptidase A) 1 antibody; angiotensin 1 converting enzyme 1; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme 1; angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1); Angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; CD143; Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme); Dipeptidyl carboxypeptidase I; dipeptidyl peptidase; Kininase II
Gene Aliases: Ace; AW208573; CD143; Dcp1; StsRR92
UniProt ID: (Mouse) P09470, (Rat) P47820
Entrez Gene ID: (Mouse) 11421, (Rat) 24310
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.