Invitrogen
Manufacturer Code:PA581554
Catalog # PIPA581554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the N-terminus of the Mouse Angiotensin Converting Enzyme 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACE; ACE1 CD143 DCP DCP1 MGC26566 MVCD3; Angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; CD143; Dipeptidyl carboxypeptidase I; dipeptidyl peptidase; Kininase II
Gene Aliases: Ace; AW208573; CD143; Dcp1
UniProt ID: (Mouse) P09470
Entrez Gene ID: (Mouse) 11421
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.