Invitrogen
Manufacturer Code:PA5111652
Catalog # PIPA5111652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human ACE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACE; ACE ACE1 Angiotensin-converting enzyme Angiotensin-converting enzyme soluble form AW208573 BOS_19238 CD143 DCP DCP1 Dipeptidyl carboxypeptidase I I79_009670 ICH Kininase II MGC26566 MVCD3 StsRR92; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; Angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; carboxycathepsin; CD143; CD143 antigen; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase I; Kininase II; peptidase P; testicular ECA
Gene Aliases: ACE; ACE1; CD143; DCP; DCP1; ICH; MVCD3
UniProt ID: (Human) P12821
Entrez Gene ID: (Human) 1636
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.