Proteintech
Manufacturer Code:247431AP150UL
Catalog # 50-173-4518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACE Fusion Protein Ag20654 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACE; ACE ACE1 Angiotensin converting enzyme CD143 DCP DCP1 Dipeptidyl carboxypeptidase I Kininase II MVCD3; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; Angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; carboxycathepsin; CD143; CD143 antigen; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase I; dipeptidyl peptidase; Kininase II; peptidase P; testicular ECA
Gene Aliases: ACE; ACE1; AW208573; CD143; DCP; DCP1; ICH; MVCD3
UniProt ID: (Human) P12821, (Mouse) P09470
Entrez Gene ID: (Human) 1636, (Mouse) 11421
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.