Novus Biologicals
Manufacturer Code:NBP267692
Catalog # NBP267692
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SN0754 |
Immunogen | Synthetic peptide within Human ACE2 aa 200-230. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: ACE-related carboxypeptidase; ACEH; ACEHangiotensin I converting enzyme 2 ACE-related carboxypeptidase angiotensin I converting enzyme (peptidyl-dipeptidase A) 2 angiotensin-converting enzyme 2 Angiotensin-converting enzyme homolog DKFZp434A014 EC 3.4.17 EC 3.4.17.23 Metalloprotease MPROT15; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; Angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; Angiotensin-converting enzyme-related carboxypeptidase; Metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2
Gene Aliases: ACE2; ACEH; UNQ868/PRO1885
UniProt ID: (Human) Q9BYF1
Entrez Gene ID: (Human) 59272
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.