R&D Systems
Manufacturer Code:AF933
Catalog # AF933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ACE-2 Gln18-Ser740 Accession # Q9BYF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACE-related carboxypeptidase; ACE2 ACEH ACEHangiotensin I converting enzyme 2 ACE-related carboxypeptidase angiotensin I converting enzyme (peptidyl-dipeptidase A) 2 angiotensin-converting enzyme 2 Angiotensin-converting enzyme homolog DKFZp434A014 EC 3.4.17 EC 3.4.17.23 Metalloprotease MPROT15; ACEH; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; Angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; Angiotensin-converting enzyme-related carboxypeptidase; Metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2
Gene Aliases: ACE2; ACEH; UNQ868/PRO1885
UniProt ID: (Human) Q9BYF1
Entrez Gene ID: (Human) 59272
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.