Invitrogen
Manufacturer Code:MA525904
Catalog # PIMA525904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A2 |
Immunogen | Full length human recombit protein of ACD produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adrenocortical dysplasia protein homolog; PIP1 PTOP TINT1 TPP1; POT1 and TIN2-interacting protein; TIN2 interacting protein 1
Gene Aliases: ACD; PIP1; PTOP; TINT1; TPP1
UniProt ID: (Human) Q96AP0
Entrez Gene ID: (Human) 65057
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.