Novus Biologicals
Manufacturer Code:NBP29201020UL
Catalog # NB025914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 140-330 of human ASIC5 (NP_059115.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid sensing (proton gated) ion channel family member 5; acid sensing ion channel family member 5; acid-sensing (proton-gated) ion channel family member 5; Acid-sensing ion channel 5; Amiloride-sensitive cation channel 5; amiloride-sensitive cation channel 5 intestinal; amiloride-sensitive cation channel 5, intestinal; amiloride-sensitive sodium channel; ASIC5; HINaC; Human intestine Na(+) channel
Gene Aliases: ACCN5; ASIC5; HINAC; INAC
UniProt ID: (Human) Q9NY37
Entrez Gene ID: (Human) 51802
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.