Novus Biologicals
Manufacturer Code:NBP222409AF647
Catalog # NB222409647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S271-44 |
Immunogen | Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: acid sensing (proton gated) ion channel 1; acid sensing ion channel 1; acid-sensing (proton-gated) ion channel 1; Acid-sensing ion channel 1; Acid-sensing ion channel 1 acid-sensing ion channel 1a protein amiloride-sensitive cation channel 2 neuronal ASIC ASIC1ASIC1A BNAC2 BNaC2ACCN2 variant 3 Brain sodium channel 2 Cation channel amiloride-sensitive neuronal 2 hBNaC2; acid-sensing ion channel 1a protein; Amiloride-sensitive cation channel 2, neuronal; ASIC1; BNaC2; Brain sodium channel 2; Cation channel, amiloride-sensitive, neuronal, 2
Gene Aliases: ACCN2; ASIC; ASIC1; BNAC2
UniProt ID: (Human) P78348
Entrez Gene ID: (Human) 41
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.