Novus Biologicals
Manufacturer Code:NBP215240
Catalog # NBP215240
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ASIC1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid sensing (proton gated) ion channel 1; acid sensing ion channel 1; acid-sensing (proton-gated) ion channel 1; Acid-sensing ion channel 1; Acid-sensing ion channel 1 acid-sensing ion channel 1a protein amiloride-sensitive cation channel 2 neuronal ASIC ASIC1ASIC1A BNAC2 BNaC2ACCN2 variant 3 Brain sodium channel 2 Cation channel amiloride-sensitive neuronal 2 hBNaC2; acid-sensing ion channel 1a protein; Amiloride-sensitive cation channel 2, neuronal; ASIC1; BNaC2; Brain sodium channel 2; Cation channel, amiloride-sensitive, neuronal, 2
Gene Aliases: ACCN2; ASIC; ASIC1; BNAC2
UniProt ID: (Human) P78348
Entrez Gene ID: (Human) 41
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.