Invitrogen
Manufacturer Code:PA539004
Catalog # PIPA539004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human ACBD6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA-binding domain-containing protein 6; acyl-Coenzyme A binding domain containing 6
Gene Aliases: 0610010G04Rik; 2610100E10Rik; ACBD6
UniProt ID: (Human) Q9BR61, (Rat) Q5RJK8, (Mouse) Q9D061
Entrez Gene ID: (Human) 84320, (Rat) 289125, (Mouse) 72482
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.