Novus Biologicals
Manufacturer Code:NBP185145
Catalog # NBP185145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:AQEEVKGAEQSDNDKKMMKKSADHKNLEVIVTNGYDKDGFVQDIQNDIHASSSLNGRSTEEVKPIDENLGQTGKSAVCIHQDINDDHVEDVTGIQHLTSDSDSEVYCDSMEQFGQEESLDSFTSNNGPFQYYLGGHSSQPM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acyl-CoA binding domain containing 5 acyl-Coenzyme A binding domain containing 5 DKFZp434A2417 endozepine-related protein KIAA1996acyl-CoA-binding domain-containing protein 5 membrane-associated diazepam binding inhibitor; Acyl-CoA-binding domain-containing protein 5; acyl-Coenzyme A binding domain containing 5; endozepine-related protein; membrane-associated diazepam binding inhibitor
Gene Aliases: ACBD5; KIAA1996
UniProt ID: (Human) Q5T8D3
Entrez Gene ID: (Human) 91452
Molecular Function: enzyme modulator transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.