Invitrogen
Manufacturer Code:PA543438
Catalog # PIPA543438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ACBD4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA-binding domain-containing protein 4; acyl-Coenzyme A binding domain containing 4
Gene Aliases: 2010009P05Rik; 2010015A21Rik; ACBD4; AI849317; HMFT0700; zgc:85611
UniProt ID: (Bovine) Q2KHT9, (Human) Q8NC06, (Rat) Q6DGF9
Entrez Gene ID: (Rabbit) 100339932, (Bovine) 515848, (Dog) 490928, (Horse) 100064116, (Pig) 100520929, (Human) 79777, (Rat) 303577, (Mouse) 67131, (Guinea pig) 100724636, (Zebrafish) 406368
Molecular Function: enzyme modulator transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.