Novus Biologicals
Manufacturer Code:NBP272148APC
Catalog # NB272148APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G2 |
Immunogen | Full length recombinant protein of human ACBD3 (NP_073572) produced in HEK293T cells. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: acyl-CoA binding domain containing 3 Acyl-CoA-binding domain-containing protein 3 acyl-Coenzyme A binding domain containing 3 GCP60Golgi phosphoprotein 1 GOCAP1Peripheral benzodiazepine receptor-associated protein PAP7 golgi complex associated protein 1 60kDa Golgi complex-associated protein 1 Golgi resident protein GCP60 GOLPH1PBR- and PKA-associated protein 7 PAP7 PBR associated protein peripherial benzodiazepine receptor associated protein PKA (RIalpha)-associated protein; Acyl-CoA-binding domain-containing protein 3; acyl-Coenzyme A binding domain containing 3; GOCAP1; golgi complex associated protein 1, 60kDa; Golgi complex-associated protein 1; Golgi phosphoprotein 1; Golgi resident protein GCP60; Golgi resident protein GCP60, N-terminally processed; GOLPH1; PBR- and PKA-associated protein 7; Peripheral benzodiazepine receptor-associated protein PAP7; PKA (RIalpha)-associated protein
Gene Aliases: ACBD3; GCP60; GOCAP1; GOLPH1; PAP7
UniProt ID: (Human) B2RB29
Entrez Gene ID: (Human) 64746
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.