Invitrogen
Manufacturer Code:PA544816
Catalog # PIPA544816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ACBD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA binding domain containing 3 GCP60 GOCAP1 GOLPH1 PAP7; Acyl-CoA-binding domain-containing protein 3; acyl-Coenzyme A binding domain containing 3; DAP; DMT1-associated protein; GOCAP1; golgi complex associated protein 1, 60 kDa; golgi complex associated protein 1, 60kDa; Golgi complex-associated protein 1; Golgi phosphoprotein 1; Golgi resident protein GCP60; Golgi resident protein GCP60, N-terminally processed; GOLPH1; neurotrophin receptor alike death domain protein; p75-like apoptosis-inducing death domain protein; PBR associated protein; PBR- and PKA-associated protein 7; Peripheral benzodiazepine receptor-associated protein PAP7; peripherial benzodiazepine receptor associated protein; PKA (RIalpha)-associated protein
Gene Aliases: 60kDa; 8430407O11Rik; ACBD3; D1Ertd10e; fa20g08; GCP60; GOCAP1; GOLPH1; PAP7; si:dkey-267p15.1; wu:fa20g08; zgc:66303
UniProt ID: (Human) B2RB29, (Rat) Q7TNY6, (Mouse) Q8BMP6
Entrez Gene ID: (Horse) 100058044, (Bovine) 783375, (Rabbit) 100346815, (Human) 64746, (Rat) 289312, (Mouse) 170760, (Guinea pig) 100734765, (Zebrafish) 406270
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.