Invitrogen
Manufacturer Code:PA520829
Catalog # PIPA520829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide from near the amino terminus of human GOLPH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA-binding domain-containing protein 3; acyl-Coenzyme A binding domain containing 3; GCP60 GOCAP1 GOLPH1 PAP7 Golgi resident protein GCP60 PBR- and PKA-associated protein 7 PKA (RIalpha)-associated protein acyl-Coenzyme A binding domain containing 3 golgi complex associated protein 1 60kDa golgi phosphoprotein 1 peripheral benzodiazepine receptor-associated protein PAP7; GOCAP1; golgi complex associated protein 1, 60kDa; Golgi complex-associated protein 1; Golgi phosphoprotein 1; Golgi resident protein GCP60; Golgi resident protein GCP60, N-terminally processed; GOLPH1; PBR- and PKA-associated protein 7; Peripheral benzodiazepine receptor-associated protein PAP7; PKA (RIalpha)-associated protein
Gene Aliases: ACBD3; GCP60; GOCAP1; GOLPH1; PAP7
UniProt ID: (Human) B2RB29
Entrez Gene ID: (Human) 64746
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.