Novus Biologicals
Manufacturer Code:NBP222565AF594
Catalog # NP2226A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1500000 |
Immunogen | Recombinant human ACAT1 (34-427aa) purified from E. coli |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ACAT acetoacetyl Coenzyme A thiolase Acetoacetyl-CoA thiolase acetyl-CoA acetyltransferase 1 acetyl-CoA acetyltransferase mitochondrial acetyl-Coenzyme A acetyltransferase 1 EC 2.3.1 EC 2.3.1.9 MAT mitochondrial acetoacetyl-CoA thiolase T2 THIL; acetoacetyl Coenzyme A thiolase; Acetoacetyl-CoA thiolase; Acetyl-CoA acetyltransferase, mitochondrial; acetyl-Coenzyme A acetyltransferase 1; mitochondrial acetoacetyl-CoA thiolase; T2; testicular tissue protein Li 198
Gene Aliases: ACAT; ACAT1; MAT; T2; THIL
UniProt ID: (Human) P24752
Entrez Gene ID: (Human) 38
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.