Invitrogen
Manufacturer Code:PA316811
Catalog # APSA6811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A fusion protein construct that contains the N-terminal region of human ACAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAT1 ACAT Acetoacetyl CoA thiolase Acetoacetyl Coenzyme A thiolase Acetyl Coenzyme A acetyltransferase 1 MAT T2 THIL; acetoacetyl Coenzyme A thiolase; Acetoacetyl-CoA thiolase; acetyl-CoA acetyltransferase 1; Acetyl-CoA acetyltransferase, mitochondrial; acetyl-Coenzyme A acetyltransferase 1; mitochondrial acetoacetyl-CoA thiolase; T2; testicular tissue protein Li 198
Gene Aliases: 6330585C21Rik; ACAT; ACAT1; MAT; T2; THIL
UniProt ID: (Human) P24752, (Mouse) Q8QZT1
Entrez Gene ID: (Human) 38, (Mouse) 110446, (Chinese hamster) 100770936
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.