Abnova
Manufacturer Code:PAB18742
Catalog # 89-120-298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 257-269 at internal region of human ACAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of ACAT1.
For Research Use Only
Protein Aliases: ACAT; acetoacetyl Coenzyme A thiolase; Acetoacetyl-CoA thiolase; Acetyl-CoA acetyltransferase, mitochondrial; acetyl-Coenzyme A acetyltransferase 1; MAT; mitochondrial acetoacetyl-CoA thiolase; T2; testicular tissue protein Li 198; THIL
Gene Aliases: ACAT; ACAT1; MAT; T2; THIL
UniProt ID: (Human) P24752
Entrez Gene ID: (Human) 38
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.