Novus Biologicals
Manufacturer Code:NBP298539AF532
Catalog # NB098156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human ACAT fragment. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACACT ACACT1 ACAT-1 ACATACAT1 acyl-Coenzyme A: cholesterol acyltransferase Acyl-coenzyme A:cholesterol acyltransferase 1 Cholesterol acyltransferase 1 EC 2.3.1.26 RP11-215I23.2 STATSOAT sterol O-acyltransferase (acyl-Coenzyme A: cholesterol acyltransferase) 1 sterol O-acyltransferase 1; ACAT-1; Acyl-coenzyme A:cholesterol acyltransferase 1; Cholesterol acyltransferase 1; sterol O-acyltransferase (acyl-Coenzyme A: cholesterol acyltransferase) 1; Sterol O-acyltransferase 1
Gene Aliases: ACACT; ACACT1; ACAT; ACAT-1; ACAT1; SOAT; SOAT1; STAT
UniProt ID: (Human) P35610
Entrez Gene ID: (Human) 6646
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.