Invitrogen
Manufacturer Code:PA518209
Catalog # PIPA518209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EKLNRFQQDSQKF) corresponding to the C-terminus amino acids of ACAP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAP2 CENTB2 centaurin beta 2 CNT-B2 KIAA0041 centaurin beta2 Arf GAP with coiled coil ANK repeat and PH domains 2 ArfGAP with coiled-coil ankyrin repeat and PH domains 2; Arf GAP with coiled coil, ANK repeat and PH domains 2; Arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2; centaurin, beta 2; Centaurin-beta-2; Cnt-b2
Gene Aliases: 4832442G16; 9530039J15Rik; ACAP2; CENTB2; CNT-B2; KIAA0041; mKIAA0041
UniProt ID: (Human) Q15057, (Mouse) Q6ZQK5, (Rat) Q5FVC7
Entrez Gene ID: (Human) 23527, (Mouse) 78618, (Rat) 619382
Molecular Function: G-protein modulator enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.