Abnova
Manufacturer Code:PAB6494
Catalog # 89-132-447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human ACAP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of ACAP2.
For Research Use Only
Protein Aliases: Arf GAP with coiled coil, ANK repeat and PH domains 2; Arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2; centaurin, beta 2; Centaurin-beta-2; CENTB2; CNT-B2; KIAA0041
Gene Aliases: ACAP2; CENTB2; CNT-B2; KIAA0041
UniProt ID: (Human) Q15057
Entrez Gene ID: (Human) 23527
Molecular Function: G-protein modulator enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.