Invitrogen
Manufacturer Code:PA582655
Catalog # PIPA582655
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ACADVL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD6 LCACD VLCAD; acyl-Coenzyme A dehydrogenase, very long chain; MVLCAD; Very long chain Acyl-Coa dehydrogenase; Very long-chain specific acyl-CoA dehydrogenase, mitochondrial; VLCAD; VLCAD very-long-chain acyl-CoA dehydrogenase
Gene Aliases: ACAD6; ACADVL; LCACD; VLCAD
UniProt ID: (Human) P49748, (Rat) P45953, (Mouse) P50544
Entrez Gene ID: (Human) 37, (Rat) 25363, (Mouse) 11370
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.